Pluronic® F-127

Leverandør: Biotium

Synonymer: PEG-PPG-PEG, Pluronic® F-127

59000
59000.EA 525 NOK
59000.
Pluronic® F-127
Poloxamer 407 (Pluronic® F-127)

Pluronic® F-127 is a nonionic detergent useful for solubilizing hydrophobic dye molecules such as cell-permeable AM esters of Fura-2, Fluo-3, Indo-1, or Rhod-2 calcium indicators to facilitate cell loading. Pluronic® F-127 is usually dissolved in water or DMSO with gentle heating.


  • White solid soluble in water and DMSO


We also offer ready-to-use solutions of Pluronic® F-127, 20% in DMSO and Pluronic® F-127, 10% in H2O.


Pluronic F-127 is a registered trade mark of BASF.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082049
CAS-nummer: 9003-11-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR