Pluronic® F-127 10% water

Leverandør: Biotium

Synonymer: PEG-PPG-PEG, Pluronic® F-127

59005
59005.EA 1350 NOK
59005.
Pluronic® F-127 10% water
Poloxamer 407 (Pluronic® F-127)

Ready-to-use solution of Pluronic® F-127, a nonionic detergent useful for solubilizing hydrophobic dye molecules such as AM esters of Fura-2, Fluo-3, Indo-1, or Rhod-2 to facilitate cell loading.


We also offer Pluronic® F-127 solid and Pluronic® F-127, 20% Solution in DMSO.


Pluronic F-127 is a registered trade mark of BASF.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL nummer: MFCD00082049
CAS-nummer: 9003-11-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR