HPLC columns, SUPELCOSIL™, ABZ+Plus, Supelco®
Leverandør: Merck
59194 59196 59197 59534-U 59535-U 57917 59604 59605 59179 59194C30 59197C30 59197C40
59194.EA 11210 NOK
59194. 59196. 59197. 59534-U 59535-U 57917. 59604. 59605. 59179. 59194C30 59197C30 59197C40
HPLC columns, SUPELCOSIL™, ABZ+Plus, Supelco®
Kromatografikolonner
These phases are used to deactivate the basic compounds. Widely used in industries to analyse the difficult in basic compounds.
  • Provide short retention
  • Better peak shape
  • High efficiency
Leveringsinformasjon: Supelguard cartridge kit includes one cartridge, a stand-alone holder, a piece of tubing, and two nuts and ferrules.