Phytochemical standard, Silybin B, analytical standard, Supelco®
Leverandør: Merck
59527-5MG
59527-5MGEA 0 NOK
59527-5MG
Phytochemical standard, Silybin B, analytical standard, Supelco®
Standarder Organiske standarder
Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2abddba7253a47cbce280ee9da1c5bbbbe8c5aa&vid=b2abddba7253a47cbce280ee9da1c5bbbbe8c5aa&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Beskrivelse Silybin B
Environmentally Preferable
CAS-nr 142797-34-0
Formel C₂₅H₂₂O₁₀