Kaliumklorid, Supelco®

Leverandør: Merck

Synonymer: Potassiumchloride

M-Clarity™ quality level = MQ100

60138-250ML
60138-250MLEA 1160 NOK
60138-250ML
Kaliumklorid, Supelco®
Kaliumklorid
Formel: KCl
MW: 74.55 g/mol
Kokepunkt: 1420
Smeltepunkt: 773
Tetthet: 1.98 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8
Merck Indeks: 14,07621
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df5663a37bc9ad0f22e87cec90e6f69bd5daf0e1&vid=64199ed5df58f45462610e992e95a8ea3a2709aa&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR