Ortofosforsyre ≥85%, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Danger

6024.1000EA 1225 NOK
6024.1000 6024.2500 6024.9025 6024.9200 6024.91000PE
Ortofosforsyre ≥85%, BAKER ANALYZED® ACS, J.T.Baker®
Ortofosforsyre
Formel: H₃PO₄
Smeltepunkt: 28 °C
Tetthet: 1,71…1.87 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011340
CAS-nummer: 7664-38-2
EINECS: 231-633-2
FN: 1805
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Exceeds ACS Specifications
Assay min. 85.0 %
Alkali phosphates passes test
Color (APHA) max. 10
Chloride (Cl) max. 1 ppm
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.001 %
Nitrate (NO3) max. 5 ppm
Phosphorous or Hypophosphorous Acid passes test
Reducing Substances passes test
Sulfate (SO4) max. 0.003 %
Volatile Acids (as CH3COOH) max. 0.001 %
Trace Impurities (in ppm)
Aluminium (Al) max. 10
Arsenic (As) max. 0.5
Calcium (Ca) max. 20
Cadmium (Cd) max. 5
Copper (Cu) max. 5
Iron (Fe) max. 10
Potassium (K) max. 5
Magnesium (Mg) max. 10
Manganese (Mn) max. 0.5
Sodium (Na) max. 10
Nickel (Ni) max. 5
Lead (Pb) max. 5
Antimony (Sb) max. 4
Zinc (Zn) max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR