Human thrombin (from plasma)
Leverandør: Merck Millipore Calbiochem
605195-5000
605195-5000EA 36475 NOK
605195-5000
Human thrombin (from plasma)
Enzymer
Designed for use in thrombin clotting time tests. Key enzyme that activates factor XIII and converts fibrinogen into fibrin.