Eddiksyre glasial 99-100%, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Danger

6052.1000EA 815 NOK
6052.1000 6052.2500 6052.9025 6052.1000PE 6052.2500PE
Eddiksyre glasial 99-100%, BAKER ANALYZED® ACS, J.T.Baker®
Eddiksyre glasial (iseddik)
Formel: H₃CCOOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
FN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Exceeds ACS Specifications
Assay min. 99.7 %
Acetic anhydride ((CH3CO)2O) max. 0.01 %
Color (APHA) max. 10
Dilution Test passes test
Residue after Evaporation max. 0.001 %
Substances Reducing Dichromate passes test
Substances Reducing KMnO4 passes test
Titrable Base (meq/g) max. 0.0004
Trace Impurities (in ppm)
Aluminium (Al) max. 0.1
Arsenic (As) max. 0.05
Chloride (Cl) max. 1
Copper (Cu) max. 0.1
Iron (Fe) max. 0.2
Nickel (Ni) max. 0.1
Heavy Metals (as Pb) max. 0.5
Sulfate (SO4) max. 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR