Thymidine (2'-deoxythymidine), Millipore®

Leverandør: Merck Millipore Calbiochem
Danger

6060-5
6060-5EA 1600 NOK
6060-5
Thymidine (2'-deoxythymidine), Millipore®
Thymidine (2'-deoxythymidine)
Formel: C₁₀H₁₄N₂O₅
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006537
CAS-nummer: 50-89-5
EINECS: 200-070-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR