Silikagel, Pore size: 40 Å 35-70 mesh, high purity for column chromatography, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

60736-1KG
60736-1KGEA 1875 NOK
60736-1KG
Silikagel, Pore size: 40 Å 35-70 mesh, high purity for column chromatography, Supelco®
Silikagel
Formel: SiO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf00eb70cbc513d27a22bc71951da1c53cef6376&vid=78ea6aeed414ad7a571e246573756c7a77aad94e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR