Silikagel, Pore size: 100 Å 70-230 mesh, high purity for column chromatography, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

60745-1KG
60745-1KGEA 1650 NOK
60745-1KG
Silikagel, Pore size: 100 Å 70-230 mesh, high purity for column chromatography, Supelco®
Silikagel
Formel: SiO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011232
CAS-nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9ae04a1f80ff4d939f7709fdc025817234c7a9&vid=0b9300e6045611a50d2e5cc525da343c6e708ae7&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR