Digital stopwatch, multi-function, Spectron
Leverandør: HANHART
251.1951-00
609-0193EA 4400 NOK
609-0193
Digital stopwatch, multi-function, Spectron
Tidsur Stoppeklokker, digitale
Industrial stopwatch with two-button operation.
  • Display range: 9999,99 minutes (top line), 99999,99 minutes (bottom line)
  • Selectable: 1/100 minutes or 1/100 seconds
  • Functions at 1/100 minutes: Start/Stop/Reset; Split/Lap/Addition (Split and Lap can be read at the same time); Time of day, date; 65 memories
  • Functions if switched to 1/100 seconds: Start/Stop/Reset; Addition/Split/Lap/Short Lap (Split time can be set in advance of the actual time; Countdown (Countdown and stopwatch can be used at the same time); Time of day, date; 65 memories
Weight: 85 g