VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0197EA 860 NOK
609-0197
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
Counts down and up for 99 minutes, 59 seconds.
  • Stopwatch mode
  • Waterproof and steamproof design
  • Memory function
  • For wash-down areas, wet labs, manufacturing plants or field work
Sertifiseringer: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
Leveringsinformasjon: Supplied with battery, stand, removable suction cup and certificate of calibration.