VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0198EA 960 NOK
609-0198
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
Counts up and down for 99 minutes, 59 seconds.
  • Green LED flashes every second when time remaining is less than 10 seconds
  • Red LED indicator flashes when zero is reached, and display shows time elapsed since alarm
  • Adjustable volume or off for silent flashing LED operation
  • Three counting down memories
Sertifiseringer: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
Leveringsinformasjon: Supplied with batteries, magnetic back, stand/clip and certificate of calibration.