VWR®, Digital Timer, Two Channels
Leverandør: VWR Collection
609-0199EA 1050 NOK
609-0199
VWR®, Digital Timer, Two Channels
Tidsur Tidsur, digital
Two simultaneous countdown activities for 99 hours, 59 minutes, 59 seconds.
  • Green LED flashes while counting down
  • Red LED flashes once zero has been reached
  • Adjustable volume: High and low decibel or off for silent flashing LED operation
Sertifiseringer: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
Leveringsinformasjon: Supplied with batteries, magnetic back, clip stand and certificate of calibration.