VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0203EA 840 NOK
609-0203
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
Timing capacity: 99 minutes, 59 seconds in steps of 1 second (±0,01%).
  • LCD
  • Counts up and down
  • Numbered keys make it simple to set countdown time
  • Alarm
Sertifiseringer: Compliance with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, indicating traceability to NIST.
Leveringsinformasjon: Supplied with clip, stand, magnet, opening for lanyard, battery and certificate of calibration.