VWR®, Digital Timer, Three Channels, with Clock
Leverandør: VWR Collection
609-0204EA 855 NOK
609-0204
VWR®, Digital Timer, Three Channels, with Clock
Tidsur Tidsur, digital
Counts up and down with clock for 99 hours, 59 minutes, 59 seconds.
  • Three-line LCD with 9,6 mm high digits: Displays three different countdown/alarm times simultaneously
  • Quartz-crystal accuracy is 0,01%
  • Alarm for each channel sounds for 60 seconds
  • When zero is reached, alarm sounds, 'TIMES UP' is displayed, the timer begins counting up
  • Memory returns the display to the previously programmed time
Sertifiseringer: Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicating traceability to standards provided by NIST.More Product Information