VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0206EA 350 NOK
609-0206
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
Designed with 10 buttons, this timer allows direct input for faster programming of desired time. It consists of different mounting options, such as laying flat on a bench or for vertical view, attachment to lab coat or metal surface.
  • Counts up and down plus displays elapsed time after countdown
  • Capacity of 99 minutes, 99 seconds
  • Audible and visual alarms
  • Last time set memory
  • Requires two AAA batteries
Sertifiseringer: Individually tested and inspected using NIST in A2LA accredited laboratory (certification #2448.01), ensuring conformity to ISO 17025:20More Product Information