VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0210EA 475 NOK
609-0210
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
Counts down from 99 minutes, 59 seconds in second increments.
  • Large digits (19 mm) are readable from a distance of 6 metres
  • Three large keys makes it easy to use
  • Alarm sounds for 30 seconds and then silences automatically
  • Memory resets display to last programmed time for quick repeat of frequent, routine tests
Sertifiseringer: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standards provided by NIST.
Leveringsinformasjon: Supplied with magnet, flip-out stand, clip, battery, and certificate.