VWR®, Digital Stopwatch, Jumbo
Leverandør: VWR Collection
609-0211EA 765 NOK
609-0211
VWR®, Digital Stopwatch, Jumbo
Tidsur Stoppeklokker, digitale
Digital stopwatch with large digits (12,7 mm) for easy reading.
  • Timing capacity: 23 hours, 59 minutes, 59 seconds
  • ¹/₁₀₀ second resolution for the first 30 minutes, continues with a 1 second resolution
  • Functions: Single action, time-out, cumulative split and continuous timing
  • Clock function (time of day, AM/PM, date, day of the week, 'time of day' alarm)
  • Water-resistant, O-ring seal, and shock resistant ABS plastic case with non slip rubber sides
Sertifiseringer: Compliance with ISO/IEC 17025; accredited by A2LA indicating traceability to standardsMore Product Information