VWR®, Digital Stopwatch
Leverandør: VWR Collection
609-0213EA 1000 NOK
609-0213
VWR®, Digital Stopwatch
Tidsur Stoppeklokker, digitale
Three-line display simultaneously shows running time, cumulative split, and interval split.
  • Timing capacity: 9 hours, 59 minutes, 59 seconds and 99 hundredths
  • Functions: Single action timing, time-in/-out, continuous timing, cumulative split, interval split, countdown I and II, auto-repeat, programmable timing
  • Repeat timer provides repeat function of countdown alarm timing
  • Sixty memories recall all splits, display sample numbers 1 to 60, and show interval split longest, shortest, and average times
  • Clock function (time of day, date, day of the week, 'time