VWR®, Digital Timer, Three Channels
Leverandør: VWR Collection
62344-588
609-0092EA 660 NOK
609-0092 609-0216
VWR®, Digital Timer, Three Channels
Tidsur Tidsur, digital
Timing capacity: 19 hours, 59 minutes, 59 seconds.
  • Features: Count up/down, time-in/-out, memory for T1, T2 and T3 channels, alarm, stopwatch and ¹²/₂₄ hour clock
  • Three-line display allows simultaneous or individual countdown alarm timing of up to three tests
  • Loud, high decibel alarm sounds when time is up for 1 minute, or may be silenced manually
  • Extra large digits are easy to read from across the lab
Sertifiseringer: Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicating traceability to standards providedMore Product Information