VWR®, Digital Timer, Three Channels, with Clock
Leverandør: VWR Collection
609-0218EA 1525 NOK
609-0218
VWR®, Digital Timer, Three Channels, with Clock
Tidsur Tidsur, digital
This user-friendly bench timer displays three different times simultaneously, counts down and alarms. It also functions as a stopwatch and a 'time of day' clock displaying time to the second.
  • Timing capacity: 99 hours, 59 min, and 59 s
  • Time-of-day in 24 hour format
  • Quartz-crystal accuracy is 0,001%
  • Four rubber feet keep the timer off a wet bench
  • Three channels may be started individually or simultaneously, triple-display allows simultaneous up and down timing on different channels
  • When zero is reached, an alarm sounds, a TIME’S UP message flashes on the