VWR® Original™, Digital Timer, Three Channels
Leverandør: VWR Collection
609-0220EA 1325 NOK
609-0220
VWR® Original™, Digital Timer, Three Channels
Tidsur Tidsur, digital
This three-channel lab timer is ideal for quality control and experimental requirements. It signals drying and incubating times, helps catch all test results, and even alarms for appointments.
  • Timing capacity: 9 hours, 59 minutes, 59 seconds (resolution: 1 second)
  • Features countdown, distinct alarm and display indicator per channel
  • Alarms sound for 60 seconds; can be manually silenced or automatically silences after 1 minute
  • Extremely simple operation
  • Large 6,3 mm high LCD digits are easy to read
  • Portable thanks to small size and light weight