VWR®, Digital Timer, One Channel
Leverandør: VWR Collection
609-0224EA 1025 NOK
609-0224
VWR®, Digital Timer, One Channel
Tidsur Tidsur, digital
This single-channel timer can be programmed in countdown or count up mode.
  • Timing capacity: 99 minutes, 59 seconds
  • Resolution is 1 second
  • Alarm and memory function
  • Big digit display
Leveringsinformasjon: Supplied with magnet, clip and stand.