VWR® LP series, Precision balances
Leverandør: VWR Collection
611-2316EA 7525 NOK
611-2316 611-2315 611-2314 611-2313 611-2312 611-3349 611-3363
VWR® LP series, Precision balances
Vekter
Rugged, economical, balances that are ideal for educational establishments and laboratory use. A series that ranges from simple weighing to operation with automated internal calibration and with GLP date/time functionality. All models have a stainless steel top pan.
  • Selectable filter levels and measuring units
  • Choice of simple LCD or large graphic display (G) with added functionality for GLP and formulation
Note: Balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-86More Product Information