VWR®, Weighing Table, Granite, for Micro and Analytical Balances
Leverandør: VWR Collection
611-2549EA 32925 NOK
611-2549
VWR®, Weighing Table, Granite, for Micro and Analytical Balances
Møbler Bord
High performance micro balances and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast reading stabilisation. The VWR® weighing table features a sturdy, stable frame.
  • Granite weighing surface
  • Adjustable feet
  • Meets highest anti-vibration standards, with four vibration dampers located under the work surface
  • Facilitates fast stabilisation times
Leveringsinformasjon: Installation not included.