Infrared moisture analyser, MA160
Leverandør: Sartorius Balances
MA160-1
611-2887EA 68350 NOK
611-2887
Infrared moisture analyser, MA160
Analysatorer Fuktanalyse
Easy to use, efficient, compact moisture analysers to give fast repeatable results with an improved user interface.
  • Convenient process tracking with status light indicator
  • Rapid heat up with powerful heating elements reduces overall process time
  • BetterClean design enables the user to remove the sample chamber plate and heating module and clean in a dishwasher
  • Easy to operate with touch screen and intuitive user interface
  • Mini USB and automatic printer recognition for data communication
Designed for routine work and method development for the QCMore Product Information