Infrared moisture analyser, MA37
Leverandør: Sartorius Balances
MA37-1
611-2888EA 54700 NOK
611-2888
Infrared moisture analyser, MA37
Analysatorer Fuktanalyse
Compact moisture analyser giving fast repeatable results with an improved user interface.
  • Convenient process tracking with status light indicator
  • Rapid heat up with powerful heating elements reduces overall process time
  • BetterClean design enables the user to remove the sample chamber plate and heating module and clean in a dishwasher
  • Mini-USB and automatic printer recognition for data communication
Suitable for routine daily tasks and designed to improve the overall operational efficiency from parameter set up through the heating process to finalMore Product Information