Precision balances, GF and GX series
Leverandør: AnD Instruments Ltd (A&D)
GF-1200
611-3046EA 0 NOK
611-3046
Precision balances, GF and GX series
Vekter
High precision ranges of top pan balances from A and D featuring optimum weighing and settling time.
  • Weighing mechanism Super Hybrid Sensor (SHS) gives fast response speed - typically just one second
  • Rugged aluminium alloy enclosure and IP 54 sealed keypad
  • 14 weighing units plus counting, percentage and user programmable
  • Automatically adjusts the environmental settings to match your site for GX models
  • Built-in under hook capability and animal weighing software
Please note these products are not available in Republic of Ireland or NorthernMore Product Information