Analytical balances, Adventurer
Leverandør: OHAUS
30100620 30100622 30100621 30122610 30122657 30122611 30122612 30122613
611-3744EA 39950 NOK
611-3744 611-3791 611-3745 611-3213 611-3222 611-3214 611-3215 611-3216
Analytical balances, Adventurer
Vekter
Balances with multiple features, but uncomplicated weighing capabilities, provide the user with rapid, accurate results for routine weighing and measurement activities. Applications include weighing, formulation, check weighing and percentage weighing. Stabilisation times are typically less than three seconds.
  • Stability, accuracy and fast operation
  • Wide viewing angle colour touch screen (109 mm) for quick access to nine application modes
  • Dual-USB ports (front and rear) with GLP/GMP tracking
  • Two-piece, draft shield side doors provide good access and reduce