Static ionizer, ION-100A
Leverandør: OHAUS
30095929
611-3318EA 19100 NOK
611-3318
Static ionizer, ION-100A
Ionisatorer
Electrostatic charges on weighing containers and samples can result in false readings or instability as they create attractive and repulsive forces. The ION-100A is designed to be used in conjunction with an OHAUS balance to neutralise electrostatic charges that can lead to inaccuracies.
  • Increases the accuracy of lab measurements
  • Fan-free technology neutralises static with no sample disruption
  • Compact and versatile design; small footprint
  • Durable electrodes can be easily replaced
The ionizer neutralises the electrostatic charge between the test tubeMore Product Information