Kjeldahl analysis weighing boats, Grade 609, Whatman™
Leverandør: Whatman products (Cytiva)
10313032
611-3990EA 2950 NOK
611-3990
Kjeldahl analysis weighing boats, Grade 609, Whatman™
Skip Veieskip
Cytiva's Whatman parchment weighing boats are designed so users can collect, transfer and drop both sample and weighing boat into the acid solution during a Kjeldahl analysis without influencing analytical results.
  • A fast and reliable way to safely transfer samples for Kjeldahl analysis
  • Excellent for both weighing and quantitative sample transfer
  • Made from low nitrogen parchment paper
  • No glue or chemical additives
  • Weighing boats dissolve residue-free in the digestion solution without influencing analytical results
Cytiva's business offers weighingMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Tykkelse 0,07 mm
Vekt 80 g/m²