Infrared moisture analyser, MA35
Leverandør: Sartorius Balances
MA35M-230N
611-4357EA 36475 NOK
611-4357
Infrared moisture analyser, MA35
Analysatorer Fuktanalyse
This infrared moisture analyser is designed for use in daily routine in-process control.
  • Uniform distribution of the heat rays using an integrated reflector
  • Heating of a sample by two metal tubular-shaped heating elements
  • Uniform distribution of the heat rays using an integrated reflector
Quick and reliable analysis of the material moisture of liquid, pasty and solid substances using the thermogravimetric method.
Sertifiseringer: FDA/HACCP regulations.