VWR®, Moisture Analysers with Graphic Display, MBG Series
Leverandør: VWR Collection
611-4489EA 22250 NOK
611-4489 611-4490 611-4500 611-4517 611-4518 611-4519
VWR®, Moisture Analysers with Graphic Display, MBG Series
Analysatorer Fuktanalyse
Rugged compact moisture analysers with large backlit dot matrix graphic display for moisture determination of raw materials and finished products in the food, cosmetics, dairy, pharmaceutical, chemical agriculture, construction and ceramics industries.
  • Choice of models to meet user-requirements of capacity and precision
  • Halogen-infrared heat source
  • Double-case design to protect the weighing unit from irradiated heat
  • PT100 temperature sensor ensures process repeatability
  • Time and date information is available with printed output
  • Multi-language, customisable