VWR®, Dual-Purpose Scoops, Sterile and Non Sterile
Leverandør: VWR Collection
611-4349EA 465 NOK
611-4349 611-4350 611-4670 611-4671
VWR®, Dual-Purpose Scoops, Sterile and Non Sterile
Øser
The dual-purpose scoops are a unique economical solution that combines the function of a scoop and a weighing boat into one product. They are made from anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the scoop.
  • Ships and stores flat, and when ready to use, is easy to assemble by inserting the locking tab into the hole
  • Locking tab feature enables the scoop shape to help retain materials
  • Broad tip design keeps the scoop stable while on the scales and enables materials to be easily scooped and dispensed
  • Coated to