VWR®, Micro Anti-Static Weighing Boats
Leverandør: VWR Collection
611-5050EA 485 NOK
611-5050
VWR®, Micro Anti-Static Weighing Boats
Skip Veieskip
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material.  
  • Large tabs make boats easy to use, position and hold, especially with forceps
  • Inert material enables the boat and contents to be added to solutions to aid 100% sample recovery, where protocols allow
  • Anti-static properties prevent sample retention from static electricity
Ideal for use with pipette calibration. Smaller surface area reduces evaporation rate.