Precision balances, portable, Compass™ CX series
Leverandør: OHAUS
30428208 30428209 30428210 30428211 30428212
611-5254EA 885 NOK
611-5254 611-5255 611-5256 611-5257 611-5258
Precision balances, portable, Compass™ CX series
Vekter
Designed with energy efficiency in mind, the Compass CX series offers a battery life of up to 1000 continuous hours, allowing for uninterrupted operation. Compass™ CX scales have a stainless steel pan and are ideal for use in laboratory, industrial, education, and even household settings.
  • Streamlined two-button operation means that the balance can be operated easily
  • Large backlit LCD enables easy viewing of weighing results even in dim lighting conditions, and from a distance
  • Slim, light weight design for convenient storage allows up to four scales to be