VWR®, Weighing Boats, Sterile
Leverandør: VWR Collection
611-4672EA 1200 NOK
611-4672 611-5272 611-5273 611-5274
VWR®, Weighing Boats, Sterile
Skip Veieskip
These disposable boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Ideal for applications where there is concern of cross-contamination and 'clean' is a necessity in life science research, food, biopharmaceutical, environmental and molecular research environments. Manufactured from FDA food grade PS and packed in a cleanroom.
  • Electron beam sterilised
  • Lot number traceability for quality assurance
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases