VWR® Liquid Weighing Pour Boats, Polystyrene, White
Leverandør: VWR Collection
611-5276EA 2775 NOK
611-5276 611-5277 611-5278
VWR® Liquid Weighing Pour Boats, Polystyrene, White
Skip Veieskip
Convenient and specific tool for customers who need to weigh liquids on a scale. Deep vessel with sturdy handle designed for convenient handling and pouring of liquids.
  • Flat bottom ensures stability and prevents tipping
  • Notched pour spout design controls liquid pour
  • Operating temperature range 0 to 40 °C (104 °F)
  • Made from FDA food grade polystyrene in an ISO Class 8 clean room
  • Material biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
  • Recyclable, marked
Available in 3 sizes (50, 100 and 200 ml).