Plane weighing glass, according to Heidbrink
Leverandør: witeg Labortechnik
2.873.902
611-9090EA 2225 NOK
611-9090
Plane weighing glass, according to Heidbrink
Skip Veieskip
Borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
For all weighings in which there is an advantage when the sample is divided in thin layers.