VWR®, Pour-Boat Weighing Dishes
Leverandør: VWR Collection
611-9163EA 950 NOK
611-9163 611-9164 611-9165 611-3116 611-3117 611-3118
VWR®, Pour-Boat Weighing Dishes
Skip Veieskip
PS.
  • Flat bottom avoids tipping
  • Temperature resistant to 70 °C
  • Maximise recovery of often very expensive chemicals
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
Samples are easily dispensed with the pour spout design without flexing the dish. These flat bottomed dishes are made from 0,3 mm thick polystyrene for better strength, stability and handling.
The anti-static versions help reduce the risk of static build up on samples, and helps maximise sample transfer when weighing out products.