Weighing boats, disposable
Leverandør: SEMADENI
2514
611-9430EA 2900 NOK
611-9430
Weighing boats, disposable
Skip Veieskip
PS, square, white.
  • For rapid weighing of powders and liquids
  • Pouring can be achieved easily by compressing the weighing dishes at both ends
  • Smooth inner surface and rounded corners.