Copper(II) ionophore I, Selectophore™ ionophore, Supelco®

Leverandør: Merck

Synonymer: o-Xylylenebis(N,N-diisobutyldithiocarbamate), o-XBDiBDTC

61193-50MG
61193-50MGEA 3100 NOK
61193-50MG
Copper(II) ionophore I, Selectophore™ ionophore, Supelco®
Copper(II) ionophore I

Neutral ionophore for Cu2+-selective polymer membrane electrodes


Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₆H₄₄N₂S₄
MW: 512.90 g/mol
MDL nummer: MFCD00214160
CAS-nummer: 125769-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff1937fb0eff4e411c3b5d68ad8c8ab9ff9c8f47&vid=4e53b8fe48cc2e9f6628e1d9f3d3a84ff37429a4&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR