Insulin syringes, Omnican® 20
Leverandør: B. Braun
9161619S
612-0140EA 870 NOK
612-0140
Insulin syringes, Omnican® 20
Sprøyter Sprøyter, medisinsk
Insulin syringe made of PP/PS with round shaped cylinder with black scale.
  • Piston with double sealing ring
  • Protective caps at the ends of the syringe for sterility
  • Integral piston stops
  • Latex-free
The crystal-clear barrel of the syringe makes sure that air bubbles are easily identified.