Fullpipette, BLAUBRAND® ETERNA, 1 merke, klasse AS
Leverandør: Brand
30502 30503 30507 30512 30514 30515 30518 930514 930503 930507 USP30515 USP30512 USP30502 USP30503 USP30507 USP30514 USP30518 930512 930515 930518 930502
612-1222EA 130 NOK
612-1222 612-1223 612-1227 612-1232 612-1234 612-1235 612-1238 BRND930514 BRND930503 BRND930507 BRNDUSP30515 BRNDUSP30512 BRNDUSP30502 BRNDUSP30503 BRNDUSP30507 BRNDUSP30514 BRNDUSP30518 BRND930512 BRND930515 BRND930518 BRND930502
Fullpipette, BLAUBRAND® ETERNA, 1 merke, klasse AS
Pipetter Fullpipetter
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in ETERNA amber stain
  • Includes a batch certificate
  • Also available (on request) with an individual certificate, USP individual certificate or DAkkS calibration certificate
DIN EN ISO 648
Sertifiseringer: DIN EN ISO 648; konformitetssertifisert