Pipette stand
Leverandør: KARTELL
255
612-1812EA 600 NOK
612-1812
Pipette stand
Pipettestativ
Stand made from PP, for 16 pipettes.
  • For 8 pipettes diameter <10 mm and 8 pipettes diameter <15 mm
  • Small holes in base allow drainage
W×D×H: 200×75×150 mm