Pipette rack, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H189600000
612-1814EA 4000 NOK
612-1814
Pipette rack, SP Bel-Art
Pipettestativ
This pipette rack made from LDPE supports pipettes horizontally without pressure on fragile tips.
  • Horizontally holds 12 volumetric or graduated pipettes 1 ml and larger
  • There is 21,3 cm between plates with a 1,9 cm diameter carrying bar at the top
  • Supports pipettes longer than 22 cm (8⅝")