Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Leverandør: GILSON
F161401
612-1816EA 1550 NOK
612-1816
Pipette stand for Gilson pipettes, rotary, CARROUSEL™
Pipettestativ
This carousel stand holds up to seven single or multi channel Gilson pipettes.
  • Robust and stable due to its heavy base
  • Made of PP and PC: Strong resistance to aggressive chemicals
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives users an easy access to the pipette of their choice
  • Space saving for a a neat, tidy and organised work bench