Målepipette, BLAUBRAND®, Type 1, klasse AS, delvis tømming
Leverandør: Brand
27721 27722 27724 27725 27727 27731 27729
612-2720EA 215 NOK
612-2720 612-2721 612-2722 612-3662 612-2723 612-2725 612-2724
Målepipette, BLAUBRAND®, Type 1, klasse AS, delvis tømming
Pipetter Målepipetter
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 1: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • Includes a batch certificate
  • Also available (on request) with an individual certificate, USP individual certificate or DAkks calibration certificate
  • Only pipettes marked (*) are suitable for cotton plugs
DIN EN ISO 835
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Sertifiseringer: DIN EN ISO 835. DE-M marking.